• USA Home
  • 8.41259 - 1,4-Difluorobenzene

EMAIL THIS PAGE TO A FRIEND
8.41259 EMD Millipore

1,4-Difluorobenzene

1,4-Difluorobenzene for synthesis. CAS 540-36-3, molar mass 114.09 g/mol., for synthesis

  • CAS Number 540-36-3

  • Linear Formula C₆H₄F₂

  • Molecular Weight 114.09 g/mol

  •  EC Number 208-742-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8412590010)
solubility   1.2 g/l
density   1.166 g/cm3 (20 °C)
flash point   2 °C
mp   -13 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.167 - 1.172
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P262: Do not get in eyes, on skin, or on clothing.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 16-24/25
Keep away from sources of ignition - No smoking.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?