• USA Home
  • 8.41261 - 2,6-Difluoroaniline

EMAIL THIS PAGE TO A FRIEND
8.41261 EMD Millipore

2,6-Difluoroaniline

2,6-Difluoroaniline for synthesis. CAS 5509-65-9, molar mass 129.1 g/mol., for synthesis

Synonym: 2,6-Difluorobenzenamine

  • CAS Number 5509-65-9

  • Linear Formula C₆H₅F₂N

  • Molecular Weight 129.1 g/mol

  •  EC Number 226-853-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8412610005)
bp   152 - 154 °C (1013 hPa)
density   1.275 g/cm3 (20 °C)
flash point   43 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.273 - 1.278
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 42 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2941 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2941 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2941 , 6.1, III
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/37/38
Flammable.Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?