EMAIL THIS PAGE TO A FRIEND
8.41263 EMD Millipore

9-Nitroanthracene

9-Nitroanthracene for synthesis. CAS 602-60-8, molar mass 223.23 g/mol., for synthesis

  • CAS Number 602-60-8

  • Linear Formula C₁₄H₉NO₂

  • Molecular Weight 223.23 g/mol

  •  EC Number 210-021-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Plastic bottle (8412630001)
mp   137 - 139 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 85.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2904 20 00
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?