• USA Home
  • 8.41265 - Ethyl 2-chloropropionate

EMAIL THIS PAGE TO A FRIEND
8.41265 EMD Millipore

Ethyl 2-chloropropionate

Ethyl 2-chloropropionate for synthesis. CAS 535-13-7, molar mass 136.57 g/mol., for synthesis

Synonym: 2-Chloropropionic acid ethyl ester

  • CAS Number 535-13-7

  • Linear Formula C₅H₉ClO₂

  • Molecular Weight 136.57 g/mol

  •  EC Number 208-610-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8412650050)
vapor pressure   4.8 hPa (25 °C)
density   1.072 g/cm3 (20 °C)
flash point   38 °C
autoignition temp.   420 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.887 - 0.892
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2935 , 3, III
Declaration (transport by air) IATA-DGR 
UN 2935 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 2935 , 3, III
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/38
Flammable.Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?