• USA Home
  • 8.41266 - 2-(tert-BOC-Oxyimino)-2-phenylacetonitrile

EMAIL THIS PAGE TO A FRIEND
8.41266 EMD Millipore

2-(tert-BOC-Oxyimino)-2-phenylacetonitrile

2-(tert-BOC-Oxyimino)-2-phenylacetonitrile for synthesis. CAS 58632-95-4, EC Number 261-370-9, molar mass 246.26 g/mol., for synthesis

  • CAS Number 58632-95-4

  • Linear Formula C₁₃H₁₄N₂O₃

  • Molecular Weight 246.26 g/mol

  •  EC Number 261-370-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8412660005)
mp   88 - 90 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 86 °C
  Melting range (upper value): ≤ 89 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?