• USA Home
  • 8.41270 - 1-(2-Phenylethyl)-4-piperidonone

EMAIL THIS PAGE TO A FRIEND
8.41270 EMD Millipore

1-(2-Phenylethyl)-4-piperidonone

1-(2-Phenylethyl)-4-piperidonone for synthesis. CAS 39742-60-4, molar mass 203.28 g/mol., for synthesis

Synonym: N-Phenethyl-4-piperidone

  • CAS Number 39742-60-4

  • Linear Formula C₁₃H₁₇NO

  • Molecular Weight 203.28 g/mol

  •  EC Number 254-613-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Plastic bottle (8412700001)
mp   58 - 60 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 58 °C
  Melting range (upper value): ≤ 61 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, III
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?