• USA Home
  • 8.41275 - 4-tert-Butylcyclohexanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.41275 EMD Millipore

4-tert-Butylcyclohexanecarboxylic acid

4-tert-Butylcyclohexanecarboxylic acid (mixture of cis and trans isomers) for synthesis. CAS 5451-55-8, molar mass 184.27 g/mol., (mixture of cis and trans isomers) for synthesis

  • CAS Number 5451-55-8

  • Linear Formula C₁₁H₂₀O₂

  • Molecular Weight 184.27 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8412750010)
mp   148 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay of cis-trans-Isomers (acidimetric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 20 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?