• USA Home
  • 8.41276 - 2-Amino-4-chloro-6-methylpyrimidine

EMAIL THIS PAGE TO A FRIEND
8.41276 EMD Millipore

2-Amino-4-chloro-6-methylpyrimidine

2-Amino-4-chloro-6-methylpyrimidine for synthesis. CAS 5600-21-5, molar mass 143.58 g/mol., for synthesis

  • CAS Number 5600-21-5

  • Linear Formula C₅H₆ClN₃

  • Molecular Weight 143.58 g/mol

  •  EC Number 227-018-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8412760005)
mp   184 - 189 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 180 °C
  Melting range (upper value): ≤ 185 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 59 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?