• USA Home
  • 8.41281 - 3,5-Dimethylbenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.41281 EMD Millipore

3,5-Dimethylbenzaldehyde

3,5-Dimethylbenzaldehyde for synthesis. CAS 5779-95-3, molar mass 134.17 g/mol., for synthesis

  • CAS Number 5779-95-3

  • Linear Formula C₉H₁₀O

  • Molecular Weight 134.17 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 ml in Glass bottle (8412810001)
bp   220 - 222 °C
density   1.09 g/cm3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.002 - 1.007
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?