EMAIL THIS PAGE TO A FRIEND
8.41284 EMD Millipore

Tropinone

Tropinone for synthesis. CAS 532-24-1, pH 8 (18 g/l, H₂O, 20 °C)., for synthesis

Synonym: 3-Tropanone, 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

  • CAS Number 532-24-1

  • Linear Formula C₈H₁₃NO

  • Molecular Weight 139.2 g/mol

  •  EC Number 208-530-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8412840005)
bp   227 °C (1013 hPa)
flash point   90 °C
mp   41 - 44 °C
pH   8 (18 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 42 °C
  Melting range (upper value): ≤ 45 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2939 80 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?