• USA Home
  • 8.41287 - Dimethyl methoxycarbonylmethanephosphonate

EMAIL THIS PAGE TO A FRIEND
8.41287 EMD Millipore

Dimethyl methoxycarbonylmethanephosphonate

Dimethyl methoxycarbonylmethanephosphonate for synthesis. CAS 5927-18-4, molar mass 166.11 g/mol., for synthesis

Synonym: Phosphonoacetic acid trimethyl ester, Trimethyl phosphonoacetate

  • CAS Number 5927-18-4

  • Linear Formula C₅H₁₁O₄P

  • Molecular Weight 166.11 g/mol

  •  EC Number 227-663-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8412870025)
bp   119 - 120 °C (1.33 hPa)
density   1.263 g/cm3 (20 °C)
flash point   70 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.260 - 1.266
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?