• USA Home
  • 8.41291 - 3-Methylthiophene-2-carbaldehyde

EMAIL THIS PAGE TO A FRIEND
8.41291 EMD Millipore

3-Methylthiophene-2-carbaldehyde

3-Methylthiophene-2-carbaldehyde for synthesis. CAS 5834-16-2, molar mass 126.18 g/mol., for synthesis

  • CAS Number 5834-16-2

  • Linear Formula C₆H₆OS

  • Molecular Weight 126.18 g/mol

  •  EC Number 227-418-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8412910025)
density   1.17 g/cm3 (20 °C)
flash point   82 °C
LD 50 oral   LD50 Rat 2204 mg/kg
LD 50 dermal   LD50 Rabbit > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 89.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.180 - 1.190
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?