• USA Home
  • 8.41292 - 1,1'-Binaphthyl-2,2'-diol

EMAIL THIS PAGE TO A FRIEND
8.41292 EMD Millipore

1,1'-Binaphthyl-2,2'-diol

1,1'-Binaphthyl-2,2'-diol for synthesis. CAS 602-09-5, molar mass 286.32 g/mol., for synthesis

  • CAS Number 602-09-5

  • Linear Formula C₂₀H₁₄O₂

  • Molecular Weight 286.32 g/mol

  •  EC Number 210-014-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8412920005)
mp   215 - 218 °C
LD 50 oral   LD50 Rat 113 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 215 °C
  Melting range (upper value): ≤ 219 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2907 29 00
Hazard Statement(s) 
H301: Toxic if swallowed.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
DU3106000
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, III
Categories of danger 
toxic, irritant
R Phrase 
R 25-36/38
Toxic if swallowed.Irritating to eyes and skin.
S Phrase 
S 37-45
Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?