EMAIL THIS PAGE TO A FRIEND
8.41298-M EMD Millipore

1-Methylindole

1-Methylindole for synthesis. CAS 603-76-9, molar mass 131.18 g/mol., for synthesis

  • CAS Number 603-76-9

  • Linear Formula C₉H₉N

  • Molecular Weight 131.18 g/mol

  •  EC Number 210-057-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8412980005)
bp   101 - 103 °C (6.67 hPa)
density   1.048 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.048 - 1.053
  Identity (IR): passes test

Price and Availability

Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?