EMAIL THIS PAGE TO A FRIEND
8.41302 EMD Millipore

3-Methylthiophene

3-Methylthiophene for synthesis. CAS 616-44-4, molar mass 98.17 g/mol., for synthesis

  • CAS Number 616-44-4

  • Linear Formula C₅H₆S

  • Molecular Weight 98.17 g/mol

  •  EC Number 210-482-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8413020025)
bp   115 - 117 °C (1013 hPa)
vapor pressure   24 hPa (20 °C)
solubility   0.40 g/l
density   1.02 g/cm3 (20 °C)
flash point   11 °C
mp   -69 °C
LD 50 oral   LD50 Rat > 2000 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.019 - 1.024
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 16
Keep away from sources of ignition - No smoking.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?