• USA Home
  • 8.41304 - 4-Methoxybenzyl mercaptan

EMAIL THIS PAGE TO A FRIEND
8.41304 EMD Millipore

4-Methoxybenzyl mercaptan

4-Methoxybenzyl mercaptan for synthesis. CAS 6258-60-2, molar mass 154.23 g/mol., for synthesis

Synonym: 4-Methoxy-α-toluenethiol

  • CAS Number 6258-60-2

  • Linear Formula C₈H₁₀OS

  • Molecular Weight 154.23 g/mol

  •  EC Number 228-393-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8413040005)
bp   90 - 95 °C (0.67 hPa)
density   1.11 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.110 - 1.115
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
6
Sulfides and sulfites as well as other reducing substances, inorganic cyanides and isonitriles: Stir into sodium hypochlorite solution (Cat. No. 105614) and, if necessary, leave to react for several days. Caution: some substances may show a violent reaction! Draw off any toxic or flammable gases that may be formed. Excess oxidant should be neutralized with sodium thiosulfate (Cat. No. 106513). Container D or E.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
UN 3334 , 9, III
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?