• USA Home
  • 8.41305 - p-Tolylhydrazine hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.41305 EMD Millipore

p-Tolylhydrazine hydrochloride

p-Tolylhydrazine hydrochloride for synthesis. CAS 637-60-5, EC Number 211-295-2, chemical formula C₇H₁₀N₂*HCl., for synthesis

Synonym: 4-Methylphenylhydrazine hydrochloride

  • CAS Number 637-60-5

  • Linear Formula C₇H₁₁ClN₂

  • Molecular Weight 158.63 g/mol

  •  EC Number 211-295-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8413050005)
mp   215 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Hazard Statement(s) 
H350: May cause cancer.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled.
H317: May cause an allergic skin reaction.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P201: Obtain special instructions before use.
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, III
Categories of danger 
carcinogenic, toxic, sensitizing, dangerous for the environment
R Phrase 
R 45-23/24/25-43-50/53
May cause cancer.Also toxic by inhalation, in contact with skin and if swallowed.May cause sensitisation by skin contact.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 53-36/37-45-61
Avoid exposure - obtain special instructions before use.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?