• USA Home
  • 8.41306 - 1,3,5-Tribromobenzene

EMAIL THIS PAGE TO A FRIEND
8.41306 EMD Millipore

1,3,5-Tribromobenzene

1,3,5-Tribromobenzene for synthesis. CAS 626-39-1, molar mass 314.8 g/mol., for synthesis

  • CAS Number 626-39-1

  • Linear Formula C₆H₃Br₃

  • Molecular Weight 314.8 g/mol

  •  EC Number 210-947-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8413060025)
mp   118 - 121 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 119 °C
  Melting range (upper value): ≤ 123 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?