• USA Home
  • 8.41309 - 5-Chlorovaleronitrile

EMAIL THIS PAGE TO A FRIEND
8.41309 EMD Millipore

5-Chlorovaleronitrile

5-Chlorovaleronitrile for synthesis. CAS 6280-87-1, molar mass 117.57 g/mol., for synthesis

Synonym: 5-Chlorovalerianic acid nitrile, 5-Chloro-pentanonitrile

  • CAS Number 6280-87-1

  • Linear Formula C₅H₈ClN

  • Molecular Weight 117.57 g/mol

  •  EC Number 228-481-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8413090005)
bp   92 - 93 °C (13.3 hPa)
density   1.447 g/cm3 (20 °C)
flash point   98 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.054 - 1.059
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, II
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?