• USA Home
  • 8.41312 - 4-Chloro-3-nitroaniline

EMAIL THIS PAGE TO A FRIEND
8.41312 EMD Millipore

4-Chloro-3-nitroaniline

4-Chloro-3-nitroaniline for synthesis. CAS 635-22-3, molar mass 172.57 g/mol., for synthesis

Synonym: 5-Amino-2-chloronitrobenzene

  • CAS Number 635-22-3

  • Linear Formula C₆H₅ClN₂O₂

  • Molecular Weight 172.57 g/mol

  •  EC Number 211-231-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8413120100)
mp   102 - 105 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 102 °C
  Melting range (upper value): ≤ 105 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 42 00
Hazard Statement(s) 
H300 + H310 + H330: Fatal if swallowed, in contact with skin or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
BX1750000
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Very toxic  Very toxic
Declaration (railroad and road) ADR, RID 
UN 2237 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2237 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2237 , 6.1, III, Marine Pollutant: P
Categories of danger 
very toxic, dangerous for the environment
R Phrase 
R 26/27/28-33-51/53
Very toxic by inhalation, in contact with skin and if swallowed.Danger of cumulative effects.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 28-36/37-45-61
After contact with skin, wash immediately with plenty of soap and water.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?