• USA Home
  • 8.41314 - Methoxycarbonylmethylene-triphenylphosphorane

EMAIL THIS PAGE TO A FRIEND
8.41314 EMD Millipore

Methoxycarbonylmethylene-triphenylphosphorane

Methoxycarbonylmethylene-triphenylphosphorane for synthesis. CAS 2605-67-6, molar mass 334.36 g/mol., for synthesis

Synonym: (Triphenylphosphoranyliden)acetic acid methyl ester

  • CAS Number 2605-67-6

  • Linear Formula C₂₁H₁₉O₂P

  • Molecular Weight 334.36 g/mol

  •  EC Number 220-018-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8413140025)
mp   165 - 169 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 165 °C
  Melting range (upper value): ≤ 169 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?