• USA Home
  • 8.41326 - O-(4-Nitrobenzyl)-hydroxylammonium chloride

EMAIL THIS PAGE TO A FRIEND
8.41326 EMD Millipore

O-(4-Nitrobenzyl)-hydroxylammonium chloride

O-(4-Nitrobenzyl)-hydroxylammonium chloride for synthesis. CAS 2086-26-2, molar mass 204.61 g/mol., for synthesis

  • CAS Number 2086-26-2

  • Linear Formula C₇H₉ClN₂O₃

  • Molecular Weight 204.61 g/mol

  •  EC Number 218-228-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8413260001)
mp   216 - 217 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 96.0 %
  Melting range (lower value): ≥ 205 °C
  Melting range (upper value): ≤ 215 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?