• USA Home
  • 8.41332 - 2-Methoxybenzoyl chloride

EMAIL THIS PAGE TO A FRIEND
8.41332 EMD Millipore

2-Methoxybenzoyl chloride

2-Methoxybenzoyl chloride for synthesis. CAS 21615-34-9, molar mass 170.59 g/mol., for synthesis

Synonym: o-Anisoyl chloride, 2-Methoxybenzoic acid chloride

  • CAS Number 21615-34-9

  • Linear Formula C₈H₇ClO₂

  • Molecular Weight 170.59 g/mol

  •  EC Number 244-477-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8413320005)
bp   128 - 129 °C (10.7 hPa)
density   1.146 g/cm3 (20 °C)
flash point   84 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.264 - 1.270
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
11
Organic acid halides, anhydrides and isocyanates can be added dropwise to an excess of methanol (Cat. No. 822283) to convert them into the corresponding methyl esters or methyl carbamates. If necessary, neutralize with sodium hydroxide solution (Cat. No. 105587). Fill into container A.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 1729 , 8, II
Declaration (transport by air) IATA-DGR 
UN 1729 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 1729 , 8, II, Segregation Group: 1 (Acids)
Categories of danger 
corrosive
R Phrase 
R 34-37
Causes burns.Irritating to respiratory system.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?