• USA Home
  • 8.41334 - 3-Bromopropionitrile

EMAIL THIS PAGE TO A FRIEND
8.41334 EMD Millipore

3-Bromopropionitrile

3-Bromopropionitrile for synthesis. CAS 2417-90-5, molar mass 133.97 g/mol., for synthesis

Synonym: ß-Bromopropionitrile

  • CAS Number 2417-90-5

  • Linear Formula C₃H₄BrN

  • Molecular Weight 133.97 g/mol

  •  EC Number 219-332-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8413340025)
bp   200 °C
density   1.615 g/cm3 (20 °C)
flash point   97 °C
LD 50 oral   LD50 Rat 25 - 50 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.630 - 1.635
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H300: Fatal if swallowed.
Precautionary Statement(s) 
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
UG1050000
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, II
Categories of danger 
toxic
R Phrase 
R 25
Toxic if swallowed.
S Phrase 
S 37-45
Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?