• USA Home
  • 8.41335 - 5-Hydroxyindole-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.41335 EMD Millipore

5-Hydroxyindole-2-carboxylic acid

5-Hydroxyindole-2-carboxylic acid for synthesis. CAS 21598-06-1, molar mass 177.16 g/mol., for synthesis

  • CAS Number 21598-06-1

  • Linear Formula C₉H₇NO₃

  • Molecular Weight 177.16 g/mol

  •  EC Number 244-468-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Plastic bottle (8413350001)
mp   240.0 - 245.0 °C (decomposition)
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (acidimetric, calc. on anhydrous substance): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?