• USA Home
  • 8.41342 - 4-Nitrophenyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
8.41342 EMD Millipore

4-Nitrophenyl isothiocyanate

4-Nitrophenyl isothiocyanate for synthesis. CAS 2131-61-5, molar mass 180.19 g/mol., for synthesis

Synonym: 1-Isothiocyanato-4-nitrobenzene

  • CAS Number 2131-61-5

  • Linear Formula C₇H₄N₂O₂S

  • Molecular Weight 180.19 g/mol

  •  EC Number 218-359-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8413420010)
bp   137 - 138 °C (15 hPa)
mp   107 - 110 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 107 °C
  Melting range (upper value): ≤ 110 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P260: Do not breathe dust.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
S Phrase 
S 22
Do not breathe dust.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?