• USA Home
  • 8.41345 - 4-Morpholine-carbonylchloride

EMAIL THIS PAGE TO A FRIEND
8.41345 EMD Millipore

4-Morpholine-carbonylchloride

4-Morpholine-carbonylchloride for synthesis. CAS 15159-40-7, molar mass 149.58 g/mol., for synthesis

Synonym: N-(Chlorocarbonyl)morpholine

  • CAS Number 15159-40-7

  • Linear Formula C₅H₈ClNO₂

  • Molecular Weight 149.58 g/mol

  •  EC Number 239-213-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8413450025)
bp   121 - 124 °C (26.7 hPa)
density   1.283 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.300 - 1.306
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H351: Suspected of causing cancer.
EUH014: Reacts violently with water.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P313: IF exposed or concerned: Get medical advice/ attention.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
11
Organic acid halides, anhydrides and isocyanates can be added dropwise to an excess of methanol (Cat. No. 822283) to convert them into the corresponding methyl esters or methyl carbamates. If necessary, neutralize with sodium hydroxide solution (Cat. No. 105587). Fill into container A.
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, II
Categories of danger 
irritant, carcinogenic
R Phrase 
R 14-36/38-40
Reacts violently with water.Irritating to eyes and skin.Limited evidence of a carcinogenic effect.
S Phrase 
S 26-30-36-38
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Never add water to this product.Wear suitable protective clothing.In case of insufficient ventilation, wear suitable respiratory equipment.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?