• USA Home
  • 8.41349 - 2'-Methoxyacetophenone

EMAIL THIS PAGE TO A FRIEND
8.41349 EMD Millipore

2'-Methoxyacetophenone

2'-Methoxyacetophenone for synthesis. CAS 579-74-8, molar mass 150.18 g/mol., for synthesis

Synonym: 2-Acetanisole

  • CAS Number 579-74-8

  • Linear Formula C₉H₁₀O₂

  • Molecular Weight 150.18 g/mol

  •  EC Number 209-446-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8413490025)
bp   119 - 120 °C (13.3 hPa)
density   1.09 g/cm3 (25 °C)
flash point   108 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.086 - 1.091
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 50 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?