EMAIL THIS PAGE TO A FRIEND
8.41350 EMD Millipore

2-Iodoanisole

2-Iodoanisole for synthesis. CAS 529-28-2, molar mass 234.03 g/mol., for synthesis

Synonym: 1-Iodo-2-methoxy-benzene, 2-Iodophenol methyl ether

  • CAS Number 529-28-2

  • Linear Formula C₇H₇IO

  • Molecular Weight 234.03 g/mol

  •  EC Number 208-456-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8413500010)
bp   240 - 242 °C
density   1.8 g/cm3 (20 °C)
flash point   >110 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.797 - 1.802
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?