• USA Home
  • 8.41357 - 1,3,5-Trimethoxybenzene

EMAIL THIS PAGE TO A FRIEND
8.41357 EMD Millipore

1,3,5-Trimethoxybenzene

1,3,5-Trimethoxybenzene for synthesis. CAS 621-23-8, molar mass 168.19 g/mol., for synthesis

Synonym: Phloroglucinol trimethyl ether

  • CAS Number 621-23-8

  • Linear Formula C₉H₁₂O₃

  • Molecular Weight 168.19 g/mol

  •  EC Number 210-673-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8413570010)
flash point   85 °C
mp   52 - 54 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 51 °C
  Melting range (upper value): ≤ 53 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?