• USA Home
  • 8.41358 - 3-Phenylpropionitrile

EMAIL THIS PAGE TO A FRIEND
8.41358 EMD Millipore

3-Phenylpropionitrile

3-Phenylpropionitrile for synthesis. CAS 645-59-0, molar mass 131.17 g/mol., for synthesis

Synonym: Hydrocinnamonitrile, Benzylacetonitrile

  • CAS Number 645-59-0

  • Linear Formula C₉H₉N

  • Molecular Weight 131.17 g/mol

  •  EC Number 211-447-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8413580010)
bp   261 °C
solubility   1.68 g/l (calculated)
density   1.001 g/cm3 (20 °C)
flash point   110 °C
mp   -2 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.997 - 1.002
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H301: Toxic if swallowed.
Precautionary Statement(s) 
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, III
Categories of danger 
toxic
R Phrase 
R 25
Toxic if swallowed.
S Phrase 
S 37-45
Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?