EMAIL THIS PAGE TO A FRIEND
8.41360 EMD Millipore

o-Tolylacetic acid

o-Tolylacetic acid for synthesis. CAS 644-36-0, molar mass 150.17 g/mol., for synthesis

Synonym: 2-Methylphenylacetic acid

  • CAS Number 644-36-0

  • Linear Formula C₉H₁₀O₂

  • Molecular Weight 150.17 g/mol

  •  EC Number 211-416-9

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   5 g in Plastic bottle (8413600005)
mp   86 - 87 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 82 °C
  Melting range (upper value): ≤ 84 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?