EMAIL THIS PAGE TO A FRIEND
8.41361 EMD Millipore

1,7-Heptanediol

1,7-Heptanediol for synthesis. CAS 629-30-1, molar mass 132.2 g/mol., for synthesis

Synonym: Heptamethylene glycol

  • CAS Number 629-30-1

  • Linear Formula C₇H₁₆O₂

  • Molecular Weight 132.2 g/mol

  •  EC Number 211-085-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8413610005)
bp   258 - 260 °C
density   0.953 g/cm3 (20 °C)
flash point   >110 °C
mp   17 - 18 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.955 - 0.962
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 39 95
RTECS 
MI9804000
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?