• USA Home
  • 8.41426 - Fluorene-2-carbaldehyde

EMAIL THIS PAGE TO A FRIEND
8.41426 EMD Millipore

Fluorene-2-carbaldehyde

Fluorene-2-carbaldehyde for synthesis. CAS 30084-90-3, molar mass 194.23 g/mol., for synthesis

  • CAS Number 30084-90-3

  • Linear Formula C₁₄H₁₀O

  • Molecular Weight 194.23 g/mol

  •  EC Number 250-035-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8414260005)
mp   86 - 87 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 82 °C
  Melting range (upper value): ≤ 84 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?