• USA Home
  • 8.41431 - 4-(Trifluoromethoxy)-aniline

EMAIL THIS PAGE TO A FRIEND
8.41431 EMD Millipore

4-(Trifluoromethoxy)-aniline

4-(Trifluoromethoxy)-aniline for synthesis. CAS 461-82-5, molar mass 177.12 g/mol., for synthesis

Synonym: α,α,α-Trifluoro-p-anisidine

  • CAS Number 461-82-5

  • Linear Formula C₇H₆F₃NO

  • Molecular Weight 177.12 g/mol

  •  EC Number 207-317-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8414310005)
bp   73 - 75 °C (13.3 hPa)
density   1.31 g/cm3 (20 °C)
flash point   80 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.321 - 1.327
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 29 00
Hazard Statement(s) 
H301: Toxic if swallowed.
H310: Fatal in contact with skin.
H315: Causes skin irritation.
H318: Causes serious eye damage.
Precautionary Statement(s) 
P280: Wear eye protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, II
R Phrase 
R 24-25-38-41
Toxic in contact with skin.Toxic if swallowed.Irritating to skin.Risk of serious damage to eyes.
S Phrase 
S 26-36/37-39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing and gloves.Wear eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?