• USA Home
  • 8.41432 - 2-Bromo-γ-butyrolactone

EMAIL THIS PAGE TO A FRIEND
8.41432 EMD Millipore

2-Bromo-γ-butyrolactone

2-Bromo-γ-butyrolactone for synthesis. CAS 5061-21-2, molar mass 164.98 g/mol., for synthesis

  • CAS Number 5061-21-2

  • Linear Formula C₄H₅BrO₂

  • Molecular Weight 164.98 g/mol

  •  EC Number 225-764-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8414320010)
bp   137 - 139 °C (8 hPa)
density   1.994 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.774 - 1.780
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 20 90
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?