• USA Home
  • 8.41438 - 5-Phenyl-1-pentanol

EMAIL THIS PAGE TO A FRIEND
8.41438 EMD Millipore

5-Phenyl-1-pentanol

5-Phenyl-1-pentanol for synthesis. CAS 10521-91-2, molar mass 164.24 g/mol., for synthesis

  • CAS Number 10521-91-2

  • Linear Formula C₁₁H₁₆O

  • Molecular Weight 164.24 g/mol

  •  EC Number 234-064-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 ml in Glass bottle (8414380001)
bp   113 - 114 °C (2 hPa)
density   0.975 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.968 - 0.974
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?