• USA Home
  • 8.41444 - Methyl 5-bromovalerate

EMAIL THIS PAGE TO A FRIEND
8.41444 EMD Millipore

Methyl 5-bromovalerate

Methyl 5-bromovalerate for synthesis. CAS 5454-83-1, molar mass 195.06 g/mol., for synthesis

Synonym: 5-Bromovaleric acid methyl ester, 5-Bromopentanoic acid methyl ester

  • CAS Number 5454-83-1

  • Linear Formula C₆H₁₁BrO₂

  • Molecular Weight 195.06 g/mol

  •  EC Number 226-709-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8414440005)
bp   79 - 80 °C (5.33 hPa)
density   1.363 g/cm3 (20 °C)
flash point   99 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.5 % (a/a)
  Density (d 20 °C/ 4 °C): 1.365 - 1.371
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?