• USA Home
  • 8.41497 - Bromoacetaldehyde dimethyl acetal

EMAIL THIS PAGE TO A FRIEND
8.41497 EMD Millipore

Bromoacetaldehyde dimethyl acetal

Bromoacetaldehyde dimethyl acetal (stabilized) for synthesis. CAS 7252-83-7, molar mass 169.02 g/mol., (stabilized) for synthesis

  • CAS Number 7252-83-7

  • Linear Formula C₄H₉BrO₂

  • Molecular Weight 169.02 g/mol

  •  EC Number 230-669-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   60 - 62 °C (13.3 hPa)
density   1.435 g/cm3 (20 °C)
flash point   53 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.432 - 1.437
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2911 00 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P260: Do not breathe vapour.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/38
Flammable.Irritating to eyes and skin.
S Phrase 
S 23
Do not breathe vapour.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?