• USA Home
  • 8.41500 - 4-tert-Butylcyclohexanone

EMAIL THIS PAGE TO A FRIEND
8.41500 EMD Millipore

4-tert-Butylcyclohexanone

4-tert-Butylcyclohexanone for synthesis. CAS 98-53-3, molar mass 154.25 g/mol., for synthesis

  • CAS Number 98-53-3

  • Linear Formula C₁₀H₁₈O

  • Molecular Weight 154.25 g/mol

  •  EC Number 202-678-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8415000100)
bp   113 - 116 °C (26.6 hPa)
density   0.893 g/cm3 (50 °C)
flash point   95 °C
autoignition temp.   440 °C
mp   46 - 50 °C
LD 50 oral   LD50 Rat 5000 mg/kg
LD 50 dermal   LD50 Rabbit 5000 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 46 °C
  Melting range (upper value): ≤ 50 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?