• USA Home
  • 8.41504 - 2-(4-Methoxyphenyl)ethanol

EMAIL THIS PAGE TO A FRIEND
8.41504 EMD Millipore

2-(4-Methoxyphenyl)ethanol

2-(4-Methoxyphenyl)ethanol for synthesis. CAS 702-23-8, EC Number 211-866-6, molar mass 152.19 g/mol., for synthesis

Synonym: 4-Methoxyphenethyl alcohol, Anisylethanol

  • CAS Number 702-23-8

  • Linear Formula C₉H₁₂O₂

  • Molecular Weight 152.19 g/mol

  •  EC Number 211-866-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8415040050)
bp   334 - 336 °C (1013 hPa)
flash point   >112 °C
mp   29 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 49 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?