• USA Home
  • 8.41508 - Pentafluorobenzonitrile

EMAIL THIS PAGE TO A FRIEND
8.41508 EMD Millipore

Pentafluorobenzonitrile

Pentafluorobenzonitrile for synthesis. CAS 773-82-0, molar mass 193.07 g/mol., for synthesis

  • CAS Number 773-82-0

  • Linear Formula C₇F₅N

  • Molecular Weight 193.07 g/mol

  •  EC Number 212-259-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8415080025)
density   1.566 g/cm3 (20 °C)
flash point   29 °C
mp   1.6 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.566 - 1.570
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 3275 , 6.1 (3), II
Declaration (transport by air) IATA-DGR 
UN 3275 , 6.1 (3), II
Declaration (transport by sea) IMDG-Code 
UN 3275 , 6.1 (3), II
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?