EMAIL THIS PAGE TO A FRIEND
8.41582 EMD Millipore

Chlorocyclopentane

Chlorocyclopentane for synthesis. CAS 930-28-9, molar mass 104.57 g/mol., for synthesis

Synonym: Cyclopentyl chloride

  • CAS Number 930-28-9

  • Linear Formula C₅H₉Cl

  • Molecular Weight 104.57 g/mol

  •  EC Number 213-212-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8415820100)
bp   113 - 114 °C (1013 hPa)
vapor pressure   28 hPa (20 °C)
solubility   0.8 g/l
density   1.01 g/cm3 (20 °C)
flash point   16 °C
autoignition temp.   380 °C
mp   -94 °C
LD 50 oral   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 89 80
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P223: Keep away from any possible contact with water, because of violent reaction and possible flash fire.
P262: Do not get in eyes, on skin, or on clothing.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 16-24
Keep away from sources of ignition - No smoking.Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?