• USA Home
  • 8.41587 - 1,3,5-Triisopropylbenzene

EMAIL THIS PAGE TO A FRIEND
8.41587 EMD Millipore

1,3,5-Triisopropylbenzene

1,3,5-Triisopropylbenzene for synthesis. CAS 717-74-8, molar mass 204.35 g/mol., for synthesis

  • CAS Number 717-74-8

  • Linear Formula C₁₅H₂₄

  • Molecular Weight 204.35 g/mol

  •  EC Number 211-941-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8415870050)
density   0.85 g/cm3 (20 °C)
flash point   86 °C
mp   -7 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2902 90 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?