EMAIL THIS PAGE TO A FRIEND
8.41626 EMD Millipore

Diglycolic acid

Diglycolic acid for synthesis. CAS 110-99-6, molar mass 134.09 g/mol., for synthesis

  • CAS Number 110-99-6

  • Linear Formula C₄H₆O₅

  • Molecular Weight 134.09 g/mol

  •  EC Number 203-823-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8416260100)
solubility   417 g/l
mp   144 - 145 °C
LD 50 oral   LD50 Rat 500 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 141 °C
  Melting range (upper value): ≤ 144 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
Hazard Statement(s) 
H302: Harmful if swallowed.
RTECS 
AJ 1530000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?