• USA Home
  • 8.41629 - Phenylglyoxylic acid

EMAIL THIS PAGE TO A FRIEND
8.41629 EMD Millipore

Phenylglyoxylic acid

Phenylglyoxylic acid for synthesis. CAS 611-73-4, EC Number 210-278-7, chemical formula (C₆H₅)COCOOH., for synthesis

Synonym: Benzoylformic acid

  • CAS Number 611-73-4

  • Linear Formula C₈H₆O₃

  • Molecular Weight 150.13 g/mol

  •  EC Number 210-278-7

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   10 g in Glass bottle (8416290010)
bp   147 - 151 °C (16 hPa)
density   1.38 g/cm3 (20 °C)
mp   62 - 66 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 62 °C
  Melting range (upper value): ≤ 66 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 30 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?