EMAIL THIS PAGE TO A FRIEND
8.41646 EMD Millipore

2,6-Dinitroaniline

2,6-Dinitroaniline for synthesis. CAS 606-22-4, chemical formula (NO₂)₂C₆H₃NH₂., for synthesis

  • CAS Number 606-22-4

  • Linear Formula C₆H₅N₃O₄

  • Molecular Weight 183.12 g/mol

  •  EC Number 210-108-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8416460005)
flash point   >100 °C
mp   136 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 136 °C
  Melting range (upper value): ≤ 140 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 42 00
Hazard Statement(s) 
H300 + H310 + H330: Fatal if swallowed, in contact with skin or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 2 water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Very toxic  Very toxic
Declaration (railroad and road) ADR, RID 
UN 1596 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 1596 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 1596 , 6.1, II
Categories of danger 
very toxic, dangerous for the environment
R Phrase 
R 26/27/28-33-51/53
Very toxic by inhalation, in contact with skin and if swallowed.Danger of cumulative effects.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 28-36/37-45-61
After contact with skin, wash immediately with plenty of water.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?