EMAIL THIS PAGE TO A FRIEND
8.41648 EMD Millipore

2-Decanol

2-Decanol for synthesis. CAS 1120-06-5, molar mass 158.28 g/mol., for synthesis

  • CAS Number 1120-06-5

  • Linear Formula C₁₀H₂₂O

  • Molecular Weight 158.28 g/mol

  •  EC Number 214-296-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8416480010)
bp   109 - 110 °C (13.3 hPa)
density   0.825 g/cm3 (20 °C)
flash point   85 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.5 % (a/a)
  Density (d 20 °C/ 4 °C): 0.824 - 0.828
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?