• USA Home
  • 8.41649 - 2,6-Dimethyl-4H-pyran-4-one

EMAIL THIS PAGE TO A FRIEND
8.41649 EMD Millipore

2,6-Dimethyl-4H-pyran-4-one

2,6-Dimethyl-4H-pyran-4-one for synthesis. CAS 1004-36-0, molar mass 124.13 g/mol., for synthesis

  • CAS Number 1004-36-0

  • Linear Formula C₇H₈O₂

  • Molecular Weight 124.13 g/mol

  •  EC Number 213-719-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8416490010)
mp   132 - 135 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 133 °C
  Melting range (upper value): ≤ 135 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?