• USA Home
  • 8.41657 - Cyclopropanecarboxylic acid chloride

EMAIL THIS PAGE TO A FRIEND
8.41657 EMD Millipore

Cyclopropanecarboxylic acid chloride

Cyclopropanecarboxylic acid chloride for synthesis. CAS 4023-34-1, molar mass 104.53 g/mol., for synthesis

  • CAS Number 4023-34-1

  • Linear Formula C₄H₅ClO

  • Molecular Weight 104.53 g/mol

  •  EC Number 223-684-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8416570025)
bp   120 - 121 °C (1013 hPa)
density   1.143 g/cm3 (20 °C)
flash point   23 °C
LD 50 oral   LD50 Rat 200 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.141 - 1.145
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 20 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H301: Toxic if swallowed.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P210: Keep away from heat.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
11
Organic acid halides, anhydrides and isocyanates can be added dropwise to an excess of methanol (Cat. No. 822283) to convert them into the corresponding methyl esters or methyl carbamates. If necessary, neutralize with sodium hydroxide solution (Cat. No. 105587). Fill into container A.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2920 , 8 (3), II
Declaration (transport by air) IATA-DGR 
UN 2920 , 8 (3), II
Declaration (transport by sea) IMDG-Code 
UN 2920 , 8 (3), II, Segregation Group: 1 (Acids)
Categories of danger 
flammable, toxic, corrosive
R Phrase 
R 10-25-34
Flammable.Toxic if swallowed.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?